Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-3360

Sigma-Aldrich

Sodium metasilicate nonahydrate

CP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O3Si · 9H2O
CAS Number:
Molecular Weight:
284.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

pH

12.4 (10 g/L)

SMILES string

O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

InChI

1S/2Na.O3Si.9H2O/c;;1-4(2)3;;;;;;;;;/h;;;9*1H2/q2*+1;-2;;;;;;;;;

InChI key

PHIQPXBZDGYJOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service