Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3270

Sigma-Aldrich

Sodium dodecyl sulfate

SAJ first grade, ≥95.0%

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

≤0 hPa ( 20 °C)

description

anionic detergent

Assay

≥95.0%

form

rod

mol wt

288.38 g/mol

availability

available only in Japan

pH

9.1 (10 g/L)

mp

204-207 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An-Hsun Chou et al.
Neuropharmacology, 99, 308-317 (2015-08-10)
More studies are required to develop therapeutic agents for treating spinocerebellar ataxia type 3 (SCA3), which is caused by mutant polyglutamine-expanded ataxin-3 and is the most prevalent subtype of spinocerebellar ataxias. T1-11 [N6-(4-Hydroxybenzyl) adenosine], isolated from a Chinese medicinal herb
Lei Tian et al.
Disease models & mechanisms, 8(6), 635-641 (2015-06-04)
Nemaline myopathy (NM) is one of the most common forms of congenital myopathy, and affects either fast myofibers, slow myofibers, or both. However, an animal model for congenital myopathy with fast-myofiber-specific atrophy is not available. Furthermore, mutations in the leiomodin-3
Paolo Ceppi et al.
Nature communications, 5, 5238-5238 (2014-11-05)
CD95 (APO-1/Fas) is a death receptor used by immune cells to kill cancer cells through induction of apoptosis. However, the elimination of CD95 or its ligand, CD95L, from cancer cells results in death induced by CD95R/L elimination (DICE), a type
Lan Zhang et al.
BMC cancer, 14, 885-885 (2014-11-28)
Upregulator of cell proliferation 4 (URG4) has been implicated in the oncogenesis of certain cancers. However, the correlation between URG4 expression and clinicopathological significance in human cancer remains unclear. Therefore, this study investigated its expression and clinicopathological significance in cervical
Pai-Chi Tsai et al.
Biology open, 3(8), 741-758 (2014-07-27)
Ethanol exposure during pregnancy is an established cause of birth defects, including neurodevelopmental defects. Most adult neurons are produced during the second trimester-equivalent period. The fetal neural stem cells (NSCs) that generate these neurons are an important but poorly understood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service