Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3110

Sigma-Aldrich

Sodium hypophosphite monohydrate

SAJ special grade, 82.0-86.0% anhydrous basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO2 · H2O
CAS Number:
Molecular Weight:
105.99
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

Assay

82.0-86.0% anhydrous basis

form

crystalline

availability

available only in Japan

SMILES string

O.[Na+].[O-][PH2]=O

InChI

1S/Na.H3O2P.H2O/c;1-3-2;/h;3H2,(H,1,2);1H2/q+1;;/p-1

InChI key

PLZNPHDJGFDNRM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient large-scale radical deoxygenation in alcoholic solvents using sodium hypophosphite and a phase-transfer agent: application to erythromycins.
A E Graham et al.
The Journal of organic chemistry, 65(8), 2583-2585 (2000-05-02)
Sonia Gouault-Bironneau et al.
Organic letters, 7(26), 5909-5912 (2005-12-16)
[reaction: see text] The room-temperature radical addition of sodium hypophosphite to terminal alkynes produces the previously unknown 1-alkyl-1,1-bis-H-phosphinates in moderate yield. The reaction is initiated by R3B and air and proceeds under mild conditions in an open container. The bissodium
M M Abd El-Hady et al.
Carbohydrate polymers, 92(1), 400-406 (2012-12-12)
This research mainly deals with a novel flame-retardant and UV-protection for cellulosic fabrics using ZnO nanoparticles. We present the preparation and application of ZnO nanoparticles. The size of the prepared nanoparticles was analysed using dynamic light scattering (DLS). The application
R Wadhwa et al.
Mechanisms of ageing and development, 45(3), 277-283 (1988-11-30)
Sodium hypophosphite (1 x 10(3) microM) supplementation in the diet of Zaprionus paravittiger resulted in adaptive responses in the quantitative as well as qualitative activity of peroxidase. The changes in peroxidase were in compliance with the earlier reported sodium hypophosphite
Amy K Manocchi et al.
Langmuir : the ACS journal of surfaces and colloids, 26(5), 3670-3677 (2009-11-19)
Transition-metal nanoparticles possess unique size-dependent optical, electronic, and catalytic properties on the nanoscale, which differ significantly from their bulk properties. In particular, palladium (Pd) nanoparticles have properties applicable to a wide range of applications in catalysis and electronics. However, predictable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service