Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2930

Sigma-Aldrich

Sodium dithionite

CP

Synonym(s):

Sodium hydrosulfite, Sodium hypodisulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO2SSO2Na
CAS Number:
Molecular Weight:
174.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

powder

availability

available only in Japan

pH

7.0-9.0 (20 °C, 50 g/L)

mp

>300 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])=O

InChI

1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2

InChI key

JVBXVOWTABLYPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Self-heat. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Liu et al.
Biochemistry, 48(5), 820-826 (2009-01-15)
Three paths for obtaining crystals of reduced (II-E4Q/I-K258R) cytochrome ba(3) are described, and the structures of these are reported at approximately 2.8-3.0 A resolution. Microspectrophotometry of single crystals of Thermus ba(3) oxidase at 100 K was used to show that
Elżbieta Mieczkowska et al.
Analytical and bioanalytical chemistry, 399(9), 3293-3297 (2010-11-04)
Two ordinary green light-emitting diodes used as light emitter and detector coupled with simple voltmeter form a complete, cost-effective prototype of a photometric hemoglobinometer. The device has been optimized for cuvette assays of total hemoglobin (Hb) in diluted blood using
Todd D Gruber et al.
Journal of molecular biology, 391(2), 327-340 (2009-06-09)
Galactofuranose (Galf) residues are present in cell wall glycoconjugates of numerous pathogenic microbes. Uridine 5'-diphosphate (UDP) Galf, the biosynthetic precursor of Galf-containing glycoconjugates, is produced from UDP-galactopyranose (UDP-Galp) by the flavoenzyme UDP-galactopyranose mutase (UGM). The gene encoding UGM (glf) is
Jun Shigeto et al.
PloS one, 9(8), e105332-e105332 (2014-08-20)
Lignins are aromatic heteropolymers that arise from oxidative coupling of lignin precursors, including lignin monomers (p-coumaryl, coniferyl, and sinapyl alcohols), oligomers, and polymers. Whereas plant peroxidases have been shown to catalyze oxidative coupling of monolignols, the oxidation activity of well-studied
Baharak Bahmani et al.
Lasers in surgery and medicine, 46(7), 582-592 (2014-06-26)
Ovarian cancer remains the deadliest malignancy of the female reproductive system. The ability to identify and destroy all ovarian tumor nodules may have a termendous impact on preventing tumor recurrence, and patient survival. The objective of this study is to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service