Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2510

Sigma-Aldrich

Sodium cyanide

JIS special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCN
CAS Number:
Molecular Weight:
49.01
Beilstein:
3587243
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

vapor density

1.7 (vs air)

vapor pressure

1 mmHg ( 817 °C)

Assay

≥97.0%

availability

available only in Japan

mp

563.7 °C (lit.)

SMILES string

[Na]C#N

InChI

1S/CN.Na/c1-2;

InChI key

RTVFYQXEHKQMKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivian M Zeraik et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 166(1), 17-25 (2013-05-09)
This study examined the distribution and orientation of gill O(2) chemoreceptors in Oreochromis niloticus and their role in cardiorespiratory responses to graded hypoxia. Intact fish, and a group with the first gill arch excised (operated), were submitted to graded hypoxia
Yi Dong et al.
Biochimica et biophysica acta, 1823(2), 493-504 (2011-11-08)
It has been known that the inhibition of mitochondrial cytochrome c oxidase is one of the earliest events occurring under hypoxia and this inhibition can lead to neuronal damages. Thus, the cytochrome c oxidase inhibitor sodium cyanide (NaCN) is widely
Christos Kiritsis et al.
Carbohydrate research, 364, 8-14 (2012-11-14)
A new series of 4'-C-cyano and 4'-C-cyano-4'-deoxy pyrimidine pyranonucleosides has been designed and synthesized. Commercially available 1,2,3,4,6-penta-O-acetyl-D-mannopyranose (1) was condensed with silylated 5-fluorouracil, uracil, and thymine, respectively to afford after deacetylation 1-(α-D-mannopyranosyl)nucleosides (2a-c). Subjecting 2a-c to the sequence of specific
Ming Yang et al.
Journal of the American Chemical Society, 135(10), 3768-3771 (2013-02-27)
We report a new method for stabilizing appreciable loadings (~1 wt %) of isolated gold atoms on titania and show that these catalyze the low-temperature water-gas shift reaction. The method combines a typical gold deposition/precipitation method with UV irradiation of
Arne Wick et al.
Water research, 45(12), 3638-3652 (2011-05-17)
The sludge-water distribution of a total of 41 organic micropollutants (9 phenylurea herbicides, 11 triazines, 16 biocides and 5 UV-filters) was investigated in laboratory batch experiments with fresh secondary sludge taken from a municipal WWTP. Sorption kinetics as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service