Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-2210

Sigma-Aldrich

Sodium carbonate decahydrate

JIS special grade, ≥99.0%

Synonym(s):

Sal soda, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · 10H2O
CAS Number:
Molecular Weight:
286.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

11.0-12.0 (25 °C, 50 g/L)

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hagen Frickmann et al.
PloS one, 10(8), e0135923-e0135923 (2015-08-14)
The quality of microbiological diagnostic procedures depends on pre-analytic conditions. We compared the results of 16S rRNA gene PCR and sequencing from automatically incubated blood culture materials from tropical Ghana with the results of cultural growth after automated incubation. Real-time
Ping Guo et al.
Journal of virological methods, 183(2), 139-146 (2012-05-09)
Preclinical gene therapy studies both in vitro and in vivo require high purity preparations of adeno-associated virus (AAV). Current methods for purification of AAV entail the use of centrifugation over either a CsCl or iodixanol gradient, or the use of
Feng Gu et al.
Bioresource technology, 124, 299-305 (2012-09-20)
Green liquor consists of sodium carbonate and sodium sulfide and is readily available in any kraft mills. The green liquor pretreatment process for bioethanol production was developed for wood chips. This process uses only proven technology and equipment currently used
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service