Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1560

Sigma-Aldrich

Sodium acetate trihydrate

JIS special grade, ≥99.0%

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

solid

autoignition temp.

1112 °F

availability

available only in Japan

pH

8.5-10 (25 °C, 408 g/L)

storage temp.

15-25°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Jin et al.
Bioresource technology, 117, 148-154 (2012-05-23)
Dietzia natronolimnaea JQ-AN was isolated from industrial wastewater containing aniline. Under aerobic conditions, the JQ-AN strain degraded 87% of the aniline in a 300 mg L(-1) aniline solution after 120 h of shake flask incubation in a medium containing sodium
S Srinivasa et al.
The British journal of surgery, 100(1), 66-74 (2012-11-08)
Goal-directed fluid therapy (GDFT) has been compared with liberal fluid administration in non-optimized perioperative settings. It is not known whether GDFT is of value within an enhanced recovery protocol incorporating fluid restriction. This study evaluated GDFT under these circumstances in
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace
Kit Frederiksen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 91, 9-15 (2015-01-18)
Polymeric film-forming systems (FFSs) are potential drug delivery systems for topical application to the skin. The FFSs form thin and transparent polymeric films in situ upon solvent evaporation. Their application convenience and cosmetic attributes, superior to conventional semi-solids, may offer
Do Gyun Lee et al.
Water research, 46(13), 4226-4234 (2012-06-08)
Triclosan, a synthetic antimicrobial agent, has been considered as an emerging environmental contaminant. Here we reported a triclosan-degrading wastewater bacterial isolate, Sphingopyxis strain KCY1, capable of dechlorinating triclosan with a stoichiometric release of chloride. The stain can degrade diphenyl ether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service