Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-0400

Sigma-Aldrich

Sea sand

SAJ special grade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300

grade

SAJ special grade

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service