Skip to Content
MilliporeSigma
All Photos(1)

Documents

24-5130

Sigma-Aldrich

Potassium permanganate

JIS special grade, ≥99.3%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.3%

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

pH

7.2-9.7 (20 °C, 20 g/L)

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for heterogeneous oxidations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny Li-Ying Huang et al.
Cancer research, 75(16), 3384-3397 (2015-06-27)
GNAQ and GNA11 are heterotrimeric G protein alpha subunits, which are mutated in a mutually exclusive pattern in most cases of uveal melanoma, one of the most aggressive cancers. Here we introduce the first transgenic mouse model of uveal melanoma
Arnaud Beduneau et al.
PloS one, 4(2), e4343-e4343 (2009-02-03)
We posit that the same mononuclear phagocytes (MP) that serve as target cells and vehicles for a host of microbial infections can be used to improve diagnostics and drug delivery. We also theorize that physical and biological processes such as
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Yann-Vaï Le Bihan et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 3), 409-419 (2013-03-23)
Repressor activator protein 1 (Rap1) is an essential factor involved in transcription and telomere stability in the budding yeast Saccharomyces cerevisiae. Its interaction with DNA causes hypersensitivity to potassium permanganate, suggesting local DNA melting and/or distortion. In this study, various
Ming Yang et al.
Nature communications, 6, 6445-6445 (2015-03-12)
Clostrubin is a potent antibiotic against methicillin- and vancomycin-resistant bacteria that was isolated from a strictly anaerobic bacterium Clostridium beijerinckii in 2014. This polyphenol possesses a fully substituted arene moiety on its pentacyclic scaffold, which poses a considerable challenge for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service