Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4580

Sigma-Aldrich

Potassium hexacyanoferrate(II) trihydrate

SAJ special grade, 99.5-101.0%

Synonym(s):

Potassium ferrocyanide, Yellow prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K4Fe(CN)6 · 3H2O
CAS Number:
Molecular Weight:
422.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

Assay

99.5-101.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

8.0-10.0 (25 °C, 211 g/L)

mp

70 °C (lit.)

SMILES string

O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;

InChI key

NRYOVBQTVIWDNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao Su et al.
Nanoscale, 7(45), 19129-19135 (2015-11-03)
Herein, a facile approach for shape-controlled gold nanoparticle (AuNP) decorated thionine-MoS2 nanocomposites (AuNP-Thi-MoS2) had been reported by using the synergistic effect of MoS2 and Thi. Thi was not only used as an electrochemical indicator, but also as a reducing agent
C M Gabardo et al.
The Analyst, 140(16), 5781-5788 (2015-07-17)
Fully-integrated electro-fluidic systems with micro-/nano-scale features have a wide range of applications in lab-on-a-chip systems used for biosensing, biological sample processing, and environmental monitoring. Rapid prototyping of application-specific electro-fluidic systems is envisioned to facilitate the testing, validation, and market translation
Concha Bosch-Navarro et al.
Physical chemistry chemical physics : PCCP, 17(44), 29628-29636 (2015-10-20)
The electrochemical activity of graphene is of fundamental importance to applications from energy storage to sensing, but has proved difficult to unambiguously determine due to the challenges innate to fabricating well defined graphene electrodes free from contamination. Here, we report
Marion Raters et al.
Journal of agricultural and food chemistry, 63(25), 5930-5934 (2015-06-16)
A quick and selective analytical method was developed for the simultaneous quantitation of 2-methylimidazole, 4-methylimidazole, and 2-acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, which are known to be formed by Maillard reactions. The methodology reported here employs stable-isotope dilution analysis (SIDA) using 4-methylimidazole-d6 and [(13)C6]-2-acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole as
Kamonwad Ngamchuea et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(11), 2338-2347 (2015-06-04)
Partially blocked electrodes (PBEs) are important; many applications use non-conductive nanoparticles (NPs) to introduce new electrode functionalities. As aggregation is a problem in NP immobilization, developing an in situ method to detect aggregation is vital to characterise such modified electrodes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service