Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4460

Sigma-Aldrich

Potassium cyanide

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12
Beilstein:
3593645
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥95.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

Assay

≥95.0%

form

solid

availability

available only in Japan

pH

11.5 (20 °C, 20 g/L)

mp

634 °C (lit.)

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikhyat S Bebarta et al.
Annals of emergency medicine, 64(6), 612-619 (2014-04-22)
Hydroxocobalamin is a Food and Drug Administration-approved antidote for cyanide poisoning. Cobinamide is a potential antidote that contains 2 cyanide-binding sites. To our knowledge, no study has directly compared hydroxocobalamin with cobinamide in a severe, cyanide-toxic large-animal model. Our objective
Sónia C Correia et al.
Neurobiology of disease, 45(1), 206-218 (2011-08-23)
The current study was undertaken to address the role of mitochondrial reactive oxygen species (ROS), and hypoxia inducible factor-1 alpha (HIF-1α) signaling pathway in the protection against high glucose levels in brain endothelial and NT2 neuron-like cells. Rat brain endothelial
Xiao-Zhang Yu et al.
Journal of hazardous materials, 225-226, 190-194 (2012-05-29)
A study was conducted to investigate activities of nitrate reductase (NR) and glutamine synthetase (GS) in plants during cyanide metabolism. Young rice seedlings (Oryza sativa L. cv. XZX 45) were grown in the nutrient solutions containing KNO(3) or NH(4)Cl and
Yang-Wen-Ke Liao et al.
Planta, 235(2), 225-238 (2011-07-23)
The role of mitochondrial alternative oxidase (AOX) and the relationship between systemic AOX induction, ROS formation, and systemic plant basal defense to Tobacco mosaic virus (TMV) were investigated in tomato plants. The results showed that TMV inoculation significantly increased the
P Lawson-Smith et al.
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc, 38(4), 223-237 (2011-09-01)
Cyanide (CN) intoxication inhibits cellular oxidative metabolism and may result in brain damage. Hydroxycobalamin (OHCob) is one among other antidotes that may be used following intoxication with CN. Hyperbaric oxygen (HBO2) is recommended when supportive measures or antidotes fail. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service