24-3460
Potassium hexachloroplatinate(IV)
JIS special grade, ≥99.0%
Synonym(s):
Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride
About This Item
Recommended Products
grade
JIS special grade
Assay
≥99.0%
form
powder
reaction suitability
reagent type: catalyst
core: platinum
availability
available only in Japan
pH
3-4
mp
250 °C (dec.) (lit.)
SMILES string
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
InChI
1S/6ClH.2K.Pt/h6*1H;;;/q;;;;;;2*+1;+4/p-6
InChI key
DPAIVKJGTXERIM-UHFFFAOYSA-H
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service