Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-2580

Sigma-Aldrich

Phosphomolybdic acid hydrate

SAJ special grade

Synonym(s):

Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4] · xH2O
CAS Number:
Molecular Weight:
1825.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ special grade

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

density

1.62 g/mL at 25 °C (lit.)

SMILES string

O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.H2O.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

PDDXOPNEMCREGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A precursor to hybrid organic-inorganic based molecular batteries with applications as cation-insertion electrodes.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunho Kim et al.
Antonie van Leeuwenhoek, 108(4), 871-877 (2015-08-08)
A Gram-stain negative, motile and short rod-shaped bacterial strain, designated KF89(T), was isolated from seawater of Tong-Yeong, South of Korea. Growth occurred between 10 and 35 °C (optimum 32 °C) and at pH 6.0-9.0 (optimum pH 7.0) and at 0-7
Jae-Jin Lee et al.
Current microbiology, 69(3), 286-291 (2014-04-22)
A Gram-negative, short-rod-shaped bacterial strain with gliding motility, designated as DG5A(T), was isolated from a rice field soil in South Korea. Phylogenic analysis using 16S rRNA gene sequence of the new isolate showed that strain DG5A(T) belong to the genus
Sathiyaraj Srinivasan et al.
Antonie van Leeuwenhoek, 106(2), 271-278 (2014-05-23)
Strain SR-1(T), a Gram-positive, strictly-aerobic, short-rod shaped, non-motile bacterium, was isolated from a mountain soil collected in Seoul Women's University in South Korea. Growth occurred between 15 and 37 °C (optimum, 30 °C), at pH 6.0-9.0 (optimum, pH 7.0) and with 0-2 %
Jihee Her et al.
Current microbiology, 71(2), 235-242 (2015-04-30)
A novel bacterial strain, designated Re6(T), was isolated from forest soil collected in Campbell University, North Carolina. The cells are aerobic, Gram-positive, non-motile, and rod shaped. Growth occurred at 4-42 °C (optimum, 25 °C), pH 6-9 (optimum, pH 6), and in 0-3 %
Kellen C S Gasque et al.
Bone, 72, 137-147 (2014-11-30)
Hypophosphatasia (HPP) results from ALPL gene mutations, which lead to a deficiency of tissue-nonspecific alkaline phosphatase (TNAP), and accumulation of inorganic pyrophosphate, a potent inhibitor of mineralization that is also a natural substrate of TNAP, in the extracellular space. HPP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service