Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23-0400

Sigma-Aldrich

Olive oil

CP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

grade

CP

form

liquid

availability

available only in Japan

functional group

ester
oleic acid

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.

Quality

Highly refined; low acidity.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

617.0 °F

Flash Point(C)

325 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyu Ha Lee et al.
Archives of pharmacal research, 27(10), 1016-1019 (2004-11-24)
The chromatographic separation of a methylene chloride extract of Artemisia rubripes led to the isolation of a new sesquiterpene lactone (3), together with four known compounds, a coumarin (2) and three terpenes (1, 4, and 5). Their structures were characterized
Jie Song et al.
Journal of the American Chemical Society, 126(27), 8459-8465 (2004-07-09)
Conjugated polymers capable of responding to external stimuli by changes in optical, electrical, or electrochemical properties can be used for the construction of direct sensing devices. Polydiacetylene-based systems are attractive for sensing applications due to their colorimetric response to changes
Nikdokht Etemadi-Tajbakhsh et al.
Research in pharmaceutical sciences, 15(5), 429-436 (2021-02-26)
Dorema ammoniacum D. Don (Apiaceae family) is a perennial plant whose oleo- gum resin is used as a natural remedy for various diseases, especially chronic bronchitis, and asthma. In the present study, hydromethanolic extract of D. ammoniacum root was subjected
Pietro Picconi et al.
Bioorganic & medicinal chemistry, 25(15), 3971-3979 (2017-06-11)
A novel series of pyridyl nitrofuranyl isoxazolines were synthesized and evaluated for their antibacterial activity against multiple drug resistant (MDR) Staphylococcus strains. Compounds with piperazine linker between the pyridyl group and isoxazoline ring showed better activity when compared to compounds
Song Cai et al.
Pain, 161(11), 2551-2570 (2020-06-17)
The voltage-gated calcium channels CaV3.1-3.3 constitute the T-type subfamily, whose dysfunctions are associated with epilepsy, psychiatric disorders, and chronic pain. The unique properties of low-voltage-activation, faster inactivation, and slower deactivation of these channels support their role in modulation of cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service