Skip to Content
MilliporeSigma
All Photos(1)

Documents

21-1350

Sigma-Aldrich

Nickel(II) sulfamate tetrahydrate

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(SO3NH2)2 · 4H2O
CAS Number:
Molecular Weight:
322.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

SMILES string

O.O.O.O.[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O

InChI

1S/2H3NO3S.Ni.4H2O/c2*1-5(2,3)4;;;;;/h2*(H3,1,2,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

TXRHHNYLWVQULI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service