Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-0930

Sigma-Aldrich

Nessler′s reagent

Synonym(s):

Dipotassium tetraiodomercurate(II) in dilute sodium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2HgI4
CAS Number:
Molecular Weight:
786.40
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
liquid
Pricing and availability is not currently available.

form

liquid

reaction suitability

reagent type: catalyst
core: mercury

availability

available only in Japan

density

1.097 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].I[Hg--](I)(I)I

InChI

1S/Hg.4HI.2K/h;4*1H;;/q+2;;;;;2*+1/p-4

InChI key

OPCMAZHMYZRPID-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Suitability

for determination of ammonium salts

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1A - STOT RE 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Dragonetti et al.
Dalton transactions (Cambridge, England : 2003), 48(26), 9703-9711 (2019-04-11)
Two new heteroleptic copper(i) sensitizers bearing 6,6'-dimethyl-2,2'-bipyridine-4,4'-dibenzoic acid, to anchor the dye on the titania surface, and a π-delocalized 2-(R-phenyl)-1H-phenanthro[9,10-d]imidazole (R = NPh2 or O-hexyl) ancillary ligand were prepared and well characterized. Their performance as dyes in DSSCs is quite
Haruka Takeuchi et al.
International journal of environmental research and public health, 15(9) (2018-09-13)
This study investigated the removal characteristics of N-Nitrosamines and their precursors at three pilot-scale water reclamation plants. These plants applies different integrated membrane systems: (1) microfiltration (MF)/nanofiltration (NF)/reverse osmosis (RO) membrane; (2) sand filtration/three-stage RO; and (3) ultrafiltration (UF)/NF and
S J Lambert et al.
Acta crystallographica. Section D, Biological crystallography, 55(Pt 5), 1048-1051 (1999-04-27)
Intact histone octamers, produced by a new method quickly and in bulk, were crystallized in KCl/phosphate, and the X-ray data were analysed to 2.15 A, confirming a P65 space group. This environment preserves the high-resolution structure of the octamers and
Hao Ren et al.
Biomaterials, 35(24), 6585-6594 (2014-05-09)
The cell-cycle inhibitor flavopiridol has been shown to improve recovery from spinal cord injury in animal models. However, the systemic dose of flavopiridol has side-effects and the mechanism of action is not clear. This study aimed to develop a strategy
Thorsten Müller et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 130-138 (2015-03-15)
The effects of different manufacturing methods to induce formation of amorphous content, changes of physico-chemical characteristics of powder blends and changes of aerodynamic properties over storage time (6months) analyzed with the Next Generation Impactor (NGI) are investigated. Earlier studies have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service