Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-0130

Sigma-Aldrich

1,3,6-Naphthalenetrisulfonic acid trisodium salt

SAJ first grade

Synonym(s):

Naphthalene-1,3,6-trisulfonic acid trisodium salt, Sodium 1,3,6-naphthalenetrisulfonate tribasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H5(SO3Na)3
Molecular Weight:
434.31
UNSPSC Code:
41100000

grade

SAJ first grade

availability

available only in Japan

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service