Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-1020

Sigma-Aldrich

Manganese(II) sulfate hydrate

JIS special grade, ≥99.0%

Synonym(s):

Manganous(II) sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4·xH2O
CAS Number:
Molecular Weight:
151.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jimmy E Becerra et al.
Applied and environmental microbiology, 81(11), 3880-3888 (2015-03-31)
L-Fucose is a sugar present in human secretions as part of human milk oligosaccharides, mucins, and other glycoconjugates in the intestinal epithelium. The genome of the probiotic Lactobacillus rhamnosus GG (LGG) carries a gene cluster encoding a putative L-fucose permease
Olivier Biner et al.
PloS one, 10(12), e0143963-e0143963 (2015-12-03)
Hyaluronidases are important venom components acting as spreading factor of toxic compounds. In several studies this spreading effect was tested on vertebrate tissue. However, data about the spreading activity on invertebrates, the main prey organisms of spiders, are lacking. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service