Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0730

Sigma-Aldrich

D-(+)-Maltose monohydrate

SAJ special grade, ≥99.0%

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥99.0%

availability

available only in Japan

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Aoki et al.
Journal of bacteriology, 191(6), 1777-1786 (2009-01-07)
Contact-dependent growth inhibition (CDI) is a mechanism identified in Escherichia coli by which bacteria expressing two-partner secretion proteins encoded by cdiA and cdiB bind to BamA in the outer membranes of target cells and inhibit their growth. A third gene
Dengjun He et al.
Journal of separation science, 38(13), 2229-2237 (2015-04-16)
Trehalose, a nonreducing disaccharide, has been extensively applied to food, cosmetics, and pharmaceutical goods. The resultant solution of trehalose prepared by enzymatic methods includes high amounts of maltose. However, it is quite difficult to separate maltose and trehalose on an
Li Guo et al.
Carbohydrate polymers, 128, 154-162 (2015-05-26)
To understand the susceptibility of glutinous rice starch to digestive enzymes and its potential impact on glycemic response, enzyme kinetics and in vitro digestibility of the native and gelatinized starches were investigated. The results showed that the Km values of
Sanja Milkovska-Stamenova et al.
Journal of agricultural and food chemistry, 63(25), 5911-5919 (2015-06-06)
Glycation is a ubiquitous nonenzymatic reaction of carbonyl compounds with amino groups of peptides and proteins, resulting in the formation of advanced glycation end-products (AGEs) and thereby affecting the properties and quality of thermally processed foods. In this context, mechanisms
Elodie Alinat et al.
Electrophoresis, 36(14), 1555-1563 (2015-03-31)
This work focuses on the development of a CE method allowing, for the first time, the simultaneous separation of the underivatized first seven cellodextrin oligomers (glucose, cellobiose, cellotriose, cellotetraose, cellopentaose, cellohexaose, and celloheptaose), with a view to analyze the hydrolysates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service