Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0700

Sigma-Aldrich

Malonic acid

SAJ first grade, ≥99.0%

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein:
1751370
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

132-135 °C (dec.) (lit.)

solubility

water: 1,390 g/L at 22 °C

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Zhao et al.
Nature communications, 10(1), 2566-2566 (2019-06-14)
There is clinical and scientific interest in developing local anesthetics with prolonged durations of effect from single injections. The need for such is highlighted by the current opioid epidemic. Site 1 sodium channel blockers such as tetrodotoxin (TTX) are extremely
E A Haan et al.
European journal of pediatrics, 144(6), 567-570 (1986-04-01)
A second child with a more severe deficiency of malonyl CoA decarboxylase is described. He is mildly mentally retarded and presented with vomiting, a seizure, hypoglycaemia and mild metabolic acidosis during a urinary tract infection. The urine contained increased amounts
Matthias Bauwens et al.
PloS one, 7(6), e38428-e38428 (2012-06-22)
In vivo pH imaging has been a field of interest for molecular imaging for many years. This is especially important for determining tumor acidity, an important driving force of tumor invasion and metastasis formation, but also in the process of
Anna Shevchenko et al.
International journal of pharmaceutics, 436(1-2), 403-409 (2012-07-04)
Cocrystallization and salt formation have been shown to entail substantial promise in tailoring the physicochemical properties of drug compounds, in particular, their dissolution and hygroscopicity. In this work, we report on the preparation and comparative evaluation of a new cocrystal
Christian Bonhomme et al.
Journal of the American Chemical Society, 134(30), 12611-12628 (2012-06-29)
Strontium is an element of fundamental importance in biomedical science. Indeed, it has been demonstrated that Sr(2+) ions can promote bone growth and inhibit bone resorption. Thus, the oral administration of Sr-containing medications has been used clinically to prevent osteoporosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service