Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0460

Sigma-Aldrich

Magnesium stearate

SAJ first grade

Synonym(s):

Stearic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16CO2]2Mg
CAS Number:
Molecular Weight:
591.24
Beilstein:
3919702
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

form

solid

availability

available only in Japan

concentration

4.0-4.6% Mg

mp

200 °C (lit.)

storage temp.

15-25°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Mg]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

HQKMJHAJHXVSDF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kluk et al.
International journal of pharmaceutics, 485(1-2), 1-6 (2015-03-05)
Minitablets are a novel, multi-compartment solid drug formulation, particularly intended for children between 1 and 6 years of age. Available literature shows that even infants are capable of swallowing a single minitablet. In this study, we have explored the level
Julia Grund et al.
International journal of pharmaceutics, 469(1), 94-101 (2014-04-22)
The objective of this study was to identify and evaluate key polymer properties affecting direct compression and drug release from water-insoluble matrices. Commonly used polymers, such as Kollidon(®) SR, Eudragit(®) RS and ethyl cellulose, were characterized, formulated into tablets and
Ziyaur Rahman et al.
International journal of pharmaceutics, 475(1-2), 462-470 (2014-09-01)
Clinical performance of an amorphous solid dispersion (ASD) drug product is related to the amorphous drug content because of the greater bioavailability of this form of the drug than its crystalline form. Therefore, it is paramount to monitor the amorphous
Benoît Igne et al.
International journal of pharmaceutics, 473(1-2), 219-231 (2014-07-09)
The implementation of a blend monitoring and control method based on a process analytical technology such as near infrared spectroscopy requires the selection and optimization of numerous criteria that will affect the monitoring outputs and expected blend end-point. Using a
Song Guo et al.
Brain : a journal of neurology, 137(Pt 11), 2951-2959 (2014-08-28)
The initiating mechanisms of migraine attacks are very complex but may involve the cyclic AMP signalling pathway. It is unknown whether intracellular cyclic AMP accumulation induces migraine attacks. We investigated whether administration of cilostazol, which causes cyclic AMP accumulation, may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service