Skip to Content
MilliporeSigma
All Photos(1)

Documents

19-0370

Sigma-Aldrich

Magnesium oxide

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

reaction suitability

reagent type: catalyst
core: magnesium

availability

available only in Japan

mp

2852 °C (lit.)

storage temp.

15-25°C

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from
Xiaoyan Sun et al.
BMC plant biology, 15, 116-116 (2015-05-11)
Heat stress is a critical threat to tall fescue in transitional and warm climate zones. Identification of association between molecular markers and heat tolerance-related functional traits would promote the efficient selection of heat tolerant tall fescue cultivars. Association analysis of
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the
Sanket A Deshmukh et al.
Physical chemistry chemical physics : PCCP, 14(44), 15593-15605 (2012-10-19)
Atomic scale characterization of the structure and dynamics of confined water molecules located near the metal oxide-aqueous interface is carried out using molecular dynamics simulations. Proximity effects on water molecules (H(2)O) near a magnesium oxide surface (MgO(100)) at room temperature
Jian-Bin Shi et al.
Carbohydrate research, 359, 65-69 (2012-08-29)
This work describes the structural changes of bagasse hemicelluloses during the cooking process involving active oxygen (O(2) and H(2)O(2)) and solid alkali (MgO). The hemicelluloses obtained from the bagasse raw material, pulp, and yellow liquor were analyzed by high-performance anion-exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service