Skip to Content
MilliporeSigma
All Photos(1)

Documents

18-1130

Sigma-Aldrich

D-(−)-Fructose

SAJ special grade, ≥98.0%

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

119-122 °C (dec.) (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan Monkley et al.
PloS one, 15(12), e0243807-e0243807 (2020-12-18)
Induced pluripotent stem cells (iPSC) offer the possibility to generate diverse disease-relevant cell types, from any genetic background with the use of cellular reprogramming and directed differentiation. This provides a powerful platform for disease modeling, drug screening and cell therapeutics.
B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
Marine Zwicke et al.
Annals of botany, 116(6), 1001-1015 (2015-04-09)
Extreme climatic events such as severe droughts are expected to increase with climate change and to limit grassland perennity. The present study aimed to characterize the adaptive responses by which temperate herbaceous grassland species resist, survive and recover from a
K Shiraga et al.
Carbohydrate research, 406, 46-54 (2015-02-07)
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service