Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18-0760

Sigma-Aldrich

Lead(II) oxide

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
SAJ first grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

10 mmHg ( 0 °C)

Assay

≥98.0%

form

powder

reaction suitability

reagent type: catalyst
core: lead

availability

available only in Japan

pH

9.9 (20 °C, 100 g/L)

mp

886 °C (lit.)

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Panizza et al.
Chemosphere, 90(4), 1455-1460 (2012-10-03)
In this paper the electrocatalytic properties of Ti-Ru-Sn ternary oxide (TiRuSnO(2)), PbO(2) and boron-doped diamond (BDD) anodes have been compared for the electrochemical oxidation of a real landfill leachate from an old municipal solid waste landfill (average values of COD
Guoting Li et al.
Journal of environmental sciences (China), 23(6), 998-1003 (2011-11-10)
Beta-PbO2 electrodes were prepared by electro-deposition and characterized by scanning electron microscopy, X-ray diffraction, X-ray photoelectron spectroscopy, and linear sweep voltammetry. We confirmed pure (beta-PbO2 crystals were on the electrode and it had a high oxygen evolution potential. The photoactivity
J Berashevich et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075803-075803 (2013-01-24)
The electronic properties of polycrystalline lead oxide consisting of a network of single-crystalline α-PbO platelets and the formation of native point defects in the α-PbO crystal lattice are studied using first-principles calculations. The results suggest that the polycrystalline nature of
Lei Li et al.
Journal of hazardous materials, 203-204, 274-282 (2012-01-03)
As part of contribution for developing a green recycling process of spent lead acid battery, a nanostructural lead oxide was prepared under the present investigation in low temperature calcination of lead citrate powder. The lead citrate, the precursor for preparation
Hui Zheng et al.
Huan jing ke xue= Huanjing kexue, 33(3), 857-865 (2012-05-26)
Ti-base lead dioxide electrodes (Ti/PbO2) doped with rare earth La, Ce were prepared by the electrode position. The surface morphology and crystal structure of the electrodes was analyzed by scanning electron microscopy (SEM) and X-ray diffraction (XRD), the results showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service