Skip to Content
MilliporeSigma
All Photos(1)

Documents

18-0510

Sigma-Aldrich

Lead(II) acetate trihydrate

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: lead

availability

available only in Japan

mp

75 °C (dec.) (lit.)

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunping Li et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 225-229 (2013-03-27)
Occupational and environmental exposures to lead (Pb) are a worldwide concern. DNA methylation plays an important role in the development of Pb toxicity. Here, we try to find out the evidence to prove that the methylation of the LINE-1 promoter
Christina Sobin et al.
Toxicology letters, 220(1), 44-52 (2013-04-20)
The mechanisms by which early chronic lead (Pb) exposure alter brain development have not been identified. We examined neuroimmune system effects in C57BL/6J mice with Pb exposure, including levels that may be common among children in lower socioeconomic income environments.
Jiangfei Chen et al.
Neurotoxicology and teratology, 34(6), 581-586 (2012-09-15)
Lead is a persistent metal and commonly present in our living environment. The present study was aimed to investigate lead-induced embryonic toxicity, behavioral responses, and adult learning/memory deficit in zebrafish. Lead acetate (PbAc) induced malformations such as uninflated swim bladder
Mohammed H Badawoud et al.
Neurosciences (Riyadh, Saudi Arabia), 18(1), 52-57 (2013-01-08)
To investigate the morphological changes in the peripheral motor system of the rat induced by exposure to lead. This study was conducted at the Anatomy Department, Faculty of Medicine, King Abdulaziz University, Jeddah, Kingdom of Saudi Arabia from January 2011
Yoshinori Ikenaka et al.
Environmental toxicology and chemistry, 31(10), 2300-2305 (2012-07-24)
The Republic of Zambia is rich in mineral resources, such as zinc (Zn) and lead (Pb), and mining is a key industry in Zambia. A previous study of Pb pollution in Kabwe, one of the main mining areas, found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service