Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18-0317

Sigma-Aldrich

N-Lauroylsarcosine sodium salt

SAJ first grade, ≥96.0%

Synonym(s):

N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

description

anionic surfactant

Assay

≥96.0%

mol wt

micellar avg mol wt 600

availability

available only in Japan

aggregation number

2

technique(s)

MALDI-MS: suitable
protein purification: suitable

CMC

14.6 mM (20-25°C)

application(s)

peptide synthesis

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Anionic surfactant.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunpei Okada et al.
Methods (San Diego, Calif.), 156, 66-78 (2018-12-24)
In A-to-I RNA editing, adenosine is converted to inosine in double-stranded regions of RNAs. Inosine, an abundant epitranscriptomic mark, contributes to a wide range of biological processes by regulating gene expression post-transcriptionally. To understand the effect of A-to-I RNA editing
Alexandra Steffens et al.
PLoS biology, 13(7), e1002188-e1002188 (2015-07-03)
Members of the highly conserved class of BEACH domain containing proteins (BDCPs) have been established as broad facilitators of protein-protein interactions and membrane dynamics in the context of human diseases like albinism, bleeding diathesis, impaired cellular immunity, cancer predisposition, and
Gültekin Tamgüney et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(35), 15002-15006 (2009-08-27)
Prions are infectious proteins that cause fatal neurodegenerative diseases. Because astrocytic gliosis marked by the deposition of fibrils composed of GFAP is a prominent feature of prion disease, we asked whether GFAP might be used as a surrogate marker for
Linda A Terry et al.
Veterinary research, 42, 65-65 (2011-05-20)
Classical scrapie is a naturally transmitted prion disease of sheep and goats. Contaminated environments may contribute to the spread of disease and evidence from animal models has implicated urine, blood, saliva, placenta and faeces as possible sources of the infection.
Kunie Ando et al.
Journal of Alzheimer's disease : JAD, 40 Suppl 1, S135-S145 (2014-03-13)
Active immunization using tau phospho-peptides in tauopathy mouse models has been observed to reduce tau pathology, especially when given prior to the onset of pathology. Since tau aggregates in these models and in human tauopathies are composed of full-length tau

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service