Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-1140

Sigma-Aldrich

Iron(III) chloride hexahydrate

JIS special grade, ≥99.0%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

vapor pressure

1 mmHg ( 194 °C)

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

bp

280-285 °C (lit.)

mp

37 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Kashyap et al.
ACS nano, 8(9), 9097-9106 (2014-08-28)
Biomineralization proteins are widely used as templating agents in biomimetic synthesis of a variety of organic-inorganic nanostructures. However, the role of the protein in controlling the nucleation and growth of biomimetic particles is not well understood, because the mechanism of
Dattatraya H Dethe et al.
Organic letters, 15(3), 429-431 (2013-01-16)
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.
D C Florian Wieland et al.
Colloids and surfaces. B, Biointerfaces, 109, 74-81 (2013-04-27)
The early stages of the formation of inorganic aggregates, composed of iron compounds at the solution-air interface, were investigated in situ. The properties of the solution-air interface were changed by using different Langmuir layers. In order to get insight into
Justin D Barr et al.
Blood, 121(18), 3733-3741 (2013-01-25)
Application of ferric chloride (FeCl(3)) to exposed blood vessels is widely used to initiate thrombosis in laboratory mice. Because the mechanisms by which FeCl(3) induces endothelial injury and subsequent thrombus formation are little understood, we used scanning electron and brightfield
Ko-Lin Kuo et al.
PloS one, 7(12), e50295-e50295 (2012-12-12)
The association between intravenous (IV) iron administration and outcomes in hemodialysis (HD) patients is still debated. Therefore, this study was aimed to assess the relationship between the IV administration of ferric chloride hexahydrate (Atofen®) and cardiovascular (CV) outcome and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service