Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-1430

Sigma-Aldrich

Gold(III) chloride

SAJ special grade (dihydrate), ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AuCl3
CAS Number:
Molecular Weight:
303.33
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ special grade (dihydrate)

Assay

≥98.5%

reaction suitability

reagent type: catalyst
core: gold

availability

available only in Japan

SMILES string

Cl[Au](Cl)Cl

InChI

1S/Au.3ClH/h;3*1H/q+3;;;/p-3

InChI key

RJHLTVSLYWWTEF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed A Elzatahry et al.
International journal of nanomedicine, 7, 2829-2832 (2012-06-30)
This report concerns nanofiber composites that incorporate N-heterocyclic carbenes and the use of such composites for testing antimicrobial and antifungal activities. The nanofiber composites were produced by electrospinning mixtures of the gold chloride or gold acetate complexes of a bis(imino)acenaphthene
Yuanjing Xiao et al.
Organic letters, 14(17), 4662-4665 (2012-08-25)
The cyclic α-imino gold carbene intermediate B is most likely generated in situ via regioselective nitrene transfer from an azido group to a tethered terminal alkyne in the presence of a gold catalyst and at ambient temperature. This highly electrophilic
Martin A Prusinkiewicz et al.
The Analyst, 137(21), 4934-4942 (2012-08-18)
High spatial resolution methods to assess the physiology of growing cells should permit analysis of fungal biochemical composition. Whole colony methods cannot capture the details of physiology and organism-environment interaction, in part because the structure, function and composition of fungal
Zhi-Xiong Ma et al.
Organic letters, 14(22), 5736-5739 (2012-11-06)
An imino-Nazarov cyclization using α-aryl-substituted allenamides is described. This gold(I)-catalyzed cascade is efficient and regioselective in constructing a diverse array of synthetically useful aromatic-ring fused cyclopentenamides. The success in this transformation represents a solution to the challenge in establishing an
Vanajakshi Gudla et al.
The Journal of organic chemistry, 76(24), 9919-9933 (2011-10-28)
An intramolecular approach to generate compounds containing an arylnaphthalene lignan scaffold in high yields is presented. It involves a sequential intramolecular electrophilic attack of carbonyl on arylalkyne followed by benzannulation catalyzed by gold salt. AuCl(3) in combination with AgSbF(6) works

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service