Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-2237

Sigma-Aldrich

Dichloromethane

HPLC grade

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

HPLC grade

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1223 °F

contains

0.001-0.005% (2-methyl-2-butene)amylene as stabilizer

expl. lim.

22 %

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damian M Herz et al.
Brain : a journal of neurology, 138(Pt 6), 1658-1666 (2015-04-18)
Dopaminergic signalling in the striatum contributes to reinforcement of actions and motivational enhancement of motor vigour. Parkinson's disease leads to progressive dopaminergic denervation of the striatum, impairing the function of cortico-basal ganglia networks. While levodopa therapy alleviates basal ganglia dysfunction
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
Aria Nouri et al.
Spine, 40(14), 1092-1100 (2015-04-22)
Ambispective study. To determine whether MRI parameters improve the predictive performance of a validated clinical prediction rule used to assess functional outcomes in surgical patients with DCM. Degenerative cervical myelopathy (DCM) is the most common cause of spinal cord dysfunction
K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in
Silvia K Moreno-Forero et al.
The ISME journal, 9(1), 150-165 (2014-06-18)
The efficacy of inoculation of single pure bacterial cultures into complex microbiomes, for example, in order to achieve increased pollutant degradation rates in contaminated material (that is, bioaugmentation), has been frustrated by insufficient knowledge on the behaviour of the inoculated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service