Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-2200

Sigma-Aldrich

Dichloromethane

JIS 300, ≥99.5%, for residue analysis

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:

grade

for residue analysis

Agency

JIS 300

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.5%

autoignition temp.

1223 °F

contains

methanol as stabilizer

expl. lim.

22 %

availability

available only in Japan

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service