Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-1665

Sigma-Aldrich

Dibutyl sebacate

SAJ first grade, ≥98.0%

Synonym(s):

Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1798308
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.441 (lit.)

bp

178-179 °C/3 mmHg (lit.)

density

0.936 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

room temp

SMILES string

CCCCOC(=O)CCCCCCCCC(=O)OCCCC

InChI

1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3

InChI key

PYGXAGIECVVIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

366.8 °F - open cup

Flash Point(C)

186 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sogol Kangarlou et al.
International journal of pharmaceutics, 356(1-2), 153-166 (2008-03-22)
This research was conducted to investigate the physico-mechanical characteristics of the EC-based coating membranes plasticized with two informal ingredients of vitamin resources, cholecalciferol and alpha-tocopherol, with respect to the commercial plasticizer DBS. Proceeding the experiment, free thin polymer sheetings of
Sapna N Makhija et al.
Journal of controlled release : official journal of the Controlled Release Society, 89(1), 5-18 (2003-04-16)
A controlled porosity osmotic pump-based drug delivery system has been described in this study. Unlike the elementary osmotic pump (EOP) which consists of an osmotic core with the drug surrounded by a semipermeable membrane drilled with a delivery orifice, controlled
A Wyss et al.
Biotechnology and bioengineering, 91(2), 227-236 (2005-05-26)
The activity of penicillin acylase has been studied in aqueous and organic solvents, as free enzyme as well as immobilized within the membrane of liquid-core capsules. The activity of the enzyme is inhibited by the accumulation of the products of
E Oh et al.
International journal of pharmaceutics, 188(2), 203-219 (1999-10-16)
The surface free energy parameters of ethylcellulose (EC) films were determined using the Lifshitz-van der Waals/acid-base approach and the influence of plasticizers on their surface energetics was assessed. Films were prepared by dip-coating glass slides in organic solvents containing EC
D Serp et al.
Biotechnology and bioengineering, 82(1), 103-110 (2003-02-06)
The bioconversion of L-phenylalanine to 2-phenylethanol by Saccharomyces cerevisiae in fed-batch experiments has shown that concentrations of 2-phenylethanol of >2.9 g/L have a negative impact on the oxidative capacity of the yeast. Without tight control on ethanol production, and hence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service