Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-0690

Sigma-Aldrich

D-(+)-Glucose

SAJ first grade, ≥98.0%

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

150-152 °C (lit.)

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annelien Van Dalem et al.
Diabetologia, 58(12), 2753-2764 (2015-09-28)
We examined whether measures of glycaemic variability (GV), assessed by continuous glucose monitoring (CGM) and self-monitoring of blood glucose (SMBG), can complement or replace measures of beta cell function and insulin action in detecting the progression of preclinical disease to
A Azau et al.
Perfusion, 29(6), 496-504 (2014-03-13)
We hypothesized that the optimization of renal haemodynamics by maintaining a high level of mean arterial blood pressure (MAP) during cardiopulmonary bypass (CPB) could reduce the rate of acute kidney injury (AKI) in high-risk patients. In this randomized, controlled study
Magnus Dalén et al.
Journal of cardiovascular translational research, 8(6), 381-384 (2015-07-08)
Endomyocardial delivery in the setting of active left ventricular assist device (LVAD) support has rarely been studied. The objective was to establish a protocol for endomyocardial injections during LVAD support without compromising mechanical circulation. LVAD implantation was performed in four
Andreia V Pinho et al.
PloS one, 10(6), e0128012-e0128012 (2015-06-06)
Sirtuin 1 (Sirt1) has been reported to be a critical positive regulator of glucose-stimulated insulin secretion in pancreatic beta-cells. The effects on islet cells and blood glucose levels when Sirt1 is deleted specifically in the pancreas are still unclear. This
Mallory L Hammock et al.
Advanced materials (Deerfield Beach, Fla.), 26(35), 6138-6144 (2014-07-23)
Organic field-effect transistor (OFET) sensors can meet the need for portable and real-time diagnostics. An electronicreadout enzyme-linked immunosorbent assay using OFETs for the detection of a panel of three biomarkers in complex media to create a pre-eclampsia prognostic is demonstrated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service