Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-2340

Sigma-Aldrich

Cerium(IV) sulfate tetrahydrate

JIS special grade, 98.0-102.5%

Synonym(s):

Ceric sulfate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(SO4)2 · 4H2O
CAS Number:
Molecular Weight:
404.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

98.0-102.5%

form

solid

reaction suitability

reagent type: catalyst
core: cerium

availability

available only in Japan

pH

1.6 (20 °C, 10 g/L)

SMILES string

[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Ce.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

VZDYWEUILIUIDF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service