Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-2990

Sigma-Aldrich

Boron standard solution

1 mg/mL B, suitable for atomic absorption spectrometry, 1000 ppm B

Synonym(s):

Boric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

availability

available only in Japan

concentration

1 mg/mL B
1000 ppm B

solubility

water: soluble

density

1.440 g/cm3

suitability

suitable for atomic absorption spectrometry

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service