Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-2590

Sigma-Aldrich

Bismuth(III) nitrate pentahydrate

JIS special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
485.07
EC Number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥99.5%

form

solid

reaction suitability

reagent type: catalyst
core: bismuth

availability

available only in Japan

bp

75-80 °C (lit.)

mp

30 °C (lit.)

SMILES string

O=[N+](O[Bi](O[N+]([O-])=O)O[N+]([O-])=O)[O-].C

InChI

1S/CH4.Bi.3NO3/c;;3*2-1(3)4/h1H4;;;;/q;+3;3*-1

InChI key

DPLOIMSBRDLLRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Li et al.
Dalton transactions (Cambridge, England : 2003), 43(36), 13742-13750 (2014-08-12)
The xFe2O3/yBiOCl composites (xFe/yBi, x/y = 0/100, 5/100, 10/100, 20/100, 30/100 and 40/100 molar ratios) are prepared for the first time through an in situ hydrolysis method under hydrothermal conditions. The samples are characterized by X-ray diffraction (XRD), scanning electron
Marcela Luísa Gomes et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 20(5), 771-779 (2015-05-02)
Two bismuth(III) porphyrins-5,10,15,20-tetrakis(phenyl)porphyrinatobismuth(III) nitrate, [Bi(III)(TPP)]NO3, and the unprecedent 5,10,15,20-tetrakis(4-carbomethoxyphenyl)porphyrinatobismuth(III) nitrate, [Bi(III)(T4CMPP)]NO3, and two unprecedented antimony(V) porphyrins dichlorido(5,10,15,20-tetrakis(phenyl)porphyrinato)antimony(V) bromide, [Sb(V)(TPP)Cl2]Br, and dibromido(5,10,15,20-tetrakis(4-carbomethoxyphenyl)porphyrinato)antimony(V) bromide, [Sb(V)(T4CMPP)Br2]Br,-were synthesized by reacting the corresponding porphyrin ligand with Bi(NO3)3·5H2O or SbCl3. All compounds were characterized by UV-vis
Nahid Gharib Naseri et al.
Analytical and bioanalytical chemistry, 391(4), 1283-1292 (2008-03-21)
Catalytic adsorptive stripping voltammetry (CAdSV) has been demonstrated at a bismuth film electrode (BiFE) in an injection-moulded electrochemical micro-flow cell. The polystyrene three-electrode flow cell was fabricated with electrodes moulded from a conducting grade of polystyrene containing 40% carbon fibre
Debasish Bandyopadhyay et al.
European journal of medicinal chemistry, 89, 851-862 (2014-12-03)
Polycyclic aromatic hydrocarbons are widespread in nature with a toxicity range from non-toxic to extremely toxic. A series of pyrenyl derivatives has been synthesized following a four-step strategy where the pyrene nucleus is attached with a basic heterocyclic moiety through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service