Skip to Content
MilliporeSigma
All Photos(1)

Documents

03-1140

Sigma-Aldrich

Benzoic acid

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

≥99.5%

form

crystalline

autoignition temp.

1061 °F

availability

available only in Japan

bp

249 °C (lit.)

mp

121-125 °C (lit.)

density

1.32 g/cm3 at 20 °C

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nacht et al.
Journal of the American Academy of Dermatology, 4(1), 31-37 (1981-01-01)
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.
Deepankar Das et al.
Organic letters, 15(17), 4358-4361 (2013-08-21)
Redox-neutral formation of C-P bonds in the α-position of amines was achieved via a process that features a combination of an oxidative α-C-H bond functionalization and a reductive N-alkylation. Benzoic acid functions as an efficient catalyst in this three-component reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service