Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-0460

Sigma-Aldrich

Barium perchlorate trihydrate

JIS special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(ClO4)2 · 3H2O
CAS Number:
Molecular Weight:
390.27
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

SMILES string

O.O.O.[Ba++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/Ba.2ClHO4.3H2O/c;2*2-1(3,4)5;;;/h;2*(H,2,3,4,5);3*1H2/q+2;;;;;/p-2

InChI key

NBHDMFAHVJWOHD-UHFFFAOYSA-L

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service