Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-3780

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

SAJ first grade, ≥98.0%

Synonym(s):

β-Aminoisobutyl alcohol, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
505979
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥98.0%
bp:
165 °C (lit.)
vapor pressure:
<1 mmHg ( 25 °C)

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

<1 mmHg ( 25 °C)

Assay

≥98.0%

availability

available only in Japan

refractive index

n20/D 1.4455 (lit.)

useful pH range

9.0-10.5

pKa (25 °C)

9.7

bp

165 °C (lit.)

mp

24-28 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

SMILES string

CC(C)(N)CO

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

179.8 °F - closed cup

Flash Point(C)

82.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shia-Chung Chen et al.
Journal of hazardous materials, 179(1-3), 692-700 (2010-04-15)
In this study, the recovery of carbon dioxide using an absorbent composed of 2-amino-2-methyl-l-propanol (AMP)+monoethanolamine (MEA)+piperazine (PZ) in polytetrafluoroethylene (PTFE) membrane contactors was investigated. Experiments were conducted using various gas flow rates, liquid flow rates, absorbent blends, and pore size
K Lewandrowski et al.
Clinical chemistry, 38(11), 2286-2294 (1992-11-11)
We evaluated N-methyl-D-glucamine (MEG) as a buffer for assay of alkaline phosphatase (ALP; EC 3.1.3.1) and compared the MEG-based assay with the current International Federation of Clinical Chemistry Reference Method for ALP (IFCC/RM/ALP), in which 2-amino-2-methyl-1-propanol (AMP) is the pH
M Lamberto et al.
Analytical biochemistry, 230(2), 224-228 (1995-09-20)
The effect of derivatization with 2-amino-2-methyl-propanol on trans-3-hexadecenoic acid was investigated as part of the identification of the trans-3-hexadecenoic acid in two Nova Scotian seaweeds. After the extraction of the total fatty acids and their methylation, the monoenoic trans fraction
Vishal Govind Rao et al.
The journal of physical chemistry. B, 115(14), 3828-3837 (2011-03-23)
In this work we have shown a comparative study of changes in physicochemical properties of an aqueous solution of a common cationic surfactant cetyltrimethylammonium bromide (CTAB) upon addition of two protic ionic liquids N,N-dimethylethanolammonium [corrected] hexanoate (DAH) and N,N-dimethylethanolammonium [corrected]
Su-Hsia Lin et al.
Chemosphere, 75(10), 1410-1416 (2009-03-18)
The influence of hydrophobicity in flat-plate porous poly(vinylidene fluoride) (PVDF) and expended polytetrafluoroethylene (PTFE) membranes on CO(2) recovery using aqueous solutions of piperazine (PZ) and alkanolamine is examined. Experiments were conducted at various gas flow rates, liquid flow rates, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service