Skip to Content
MilliporeSigma
All Photos(3)

Documents

ELONGN-RO

Roche

Expand Long Range dNTPack

suitable for PCR, hotstart: no, Long Range PCR

Synonym(s):

long template pcr, longe range pcr, nucleotide mixes and sets, pcr enzymes

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
41106300
NACRES:
NA.21

usage

sufficient for ≤1000 reactions (04829069001)
sufficient for ≤200 reactions (04829042001)
sufficient for >50 reactions (04829034001)

Quality Level

feature

Long Range PCR
dNTPs included
hotstart: no

packaging

pkg of 3,500 U (04829069001 [5x700 U])
pkg of 175 U (04829034001)
pkg of 700 U (04829042001)

manufacturer/tradename

Roche

parameter

68 °C optimum reaction temp.

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

General description

The Expand Long Range dNTPack is the second-generation version of the Expand Long Template PCR System, optimized and designed to consistently amplify PCR products from 5kb up to 25kb from genomic DNA (or up to 40kb from λ DNA templates). The supplied buffer can be used for all PCR product sizes. In addition, a buffer without MgCl2, and separate vials of MgCl2 and DMSO are included if optimization is needed. Expand Long Range dNTPack contains a special enzyme mix that consists of thermostable Taq DNA Polymerase and a thermostable DNA polymerase with proofreading activity. This powerful polymerase mixture produces a high yield of PCR product. Due to the inherent 3′–5′ exonuclease activity of the proofreading polymerase, Expand Long Range dNTPack copies DNA three times more accurately than Taq DNA Polymerase.

Specificity

Heat inactivation: > 92 °C

Application

The Expand Long Range dNTPack is optimized to efficiently amplify large genomic DNA fragments from 5kb to 25kb in combination with a threefold higher fidelity than Taq DNA Polymerase. The Expand Long Range dNTPack is ideally suited for:
  • Genome mapping and sequencing
  • Contig construction
  • Characterization of cloned sequences in lambda phages or cosmids
  • Cloning and analysis of eukaryotic genes
  • Rapid identification and cloning of complete genes from genomic DNA using cDNA sequence as starting material
  • Two-step RT-PCR
Use the specially designed buffer set, ultrapure PCR Nucleotide Mix, and optimized blend of thermostable DNA polymerases for:
  • PCR
  • Large-fragment amplification
  • RT-PCR
For amplicon sizes of 20 to 35kb choose Expand 20kb PLUS PCR System, dNTPack.

Features and Benefits

Expand Long Range PCR System is ideal for genome mapping and sequencing, contig construction, characterization of cloned sequences in lambda phages or cosmids, and eukaryotic-gene cloning and analysis.
Amplify DNA fragments from 5 to 20kb from human genomic DNA, and 40kb from λDNA using supplied reaction buffers.
Use 3.75U for a standard 50μl PCR.
Each dNTPack contains 10mM additive-free sodium salt nucleotides as a ready-to-use mix.
  • Amplify long templates
up to 25kb with high specificity, yield, and increased fidelity.
  • Save time and resources.
Use one buffer, and fine-tune using the DMSO and MgCl2 solution.
  • Use this kit for all long template applications.
Obtain high yields of your desired fragment.
  • Cost-effective.
Use the convenient premixed solution of PCR grade dNTPs.

Packaging

1 kit containing 6 components

Quality

Each lot is PCR function-tested using human genomic DNA and specific primers for a 9kb, 12kb, and 15kb fragment. The enzyme mix is tested for the absence of any contaminating activities, including endo- or exonucleases and nicking activity according to the current Quality Control procedures.

Unit Definition

Volume Activity: 5 U/μl

Preparation Note

Working concentration: Standard enzyme concentration: 3.5 U per 50 μl reaction

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Use of this product is covered by US patent claims and corresponding patent claims outside the US. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patent claims require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Expand is a trademark of Roche

Kit Components Only

Product No.
Description

  • Expand Long Range Enzyme Mix

  • Expand Long Range Buffer with 12.5 mM MgCl2 5x concentrated

  • Expand Long Range Buffer without MgCl2 5x concentrated

  • MgCl2 Stock Solution 25 mM

  • DMSO 100%

  • PCR Nucleotide Mix

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwan-Wood G Lam et al.
Clinical chemistry, 58(10), 1467-1475 (2012-08-17)
A genomewide genetic and mutational profile of a fetus was recently determined via deep sequencing of maternal plasma DNA. This technology could have important applications for noninvasive prenatal diagnosis (NIPD) of many monogenic diseases. Relative haplotype dosage (RHDO) analysis, a
Jessica M Koczan et al.
Applied and environmental microbiology, 77(19), 7031-7039 (2011-08-09)
Biofilm formation plays a critical role in the pathogenesis of Erwinia amylovora and the systemic invasion of plant hosts. The functional role of the exopolysaccharides amylovoran and levan in pathogenesis and biofilm formation has been evaluated. However, the role of
Dali Huang et al.
Genes, chromosomes & cancer, 49(9), 810-818 (2010-07-08)
Chondroid lipoma, a rare benign adipose tissue tumor, may histologically resemble myxoid liposarcoma or extraskeletal myxoid chondrosarcoma, but is genetically distinct. In this study, an identical reciprocal translocation, t(11;16)(q13;p13), was identified in three chondroid lipomas, a finding consistent with previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service