Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CASPASSY-RO

Roche

Homogeneous Caspases Assay, fluorimetric

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

manufacturer/tradename

Roche

shipped in

dry ice

storage temp.

−20°C

General description

The Homogeneous Caspases Assay is a fluorimetric assay for the quantitative in vitro determination of caspases activity in microplates, which makes it especially useful for high-throughput screening.

Specificity

Enzyme activity of natural and recombinant caspase is detected by this homogeneous caspases assay. Activated caspases 2, 3, 6, 7, 8, 9, and 10 can be analyzed with different sensitivity.

Application

Specific, quantitative in vitro determination of caspases in microplates.
During apoptosis, the activation of specific proteases, called Caspases (cysteinyl-aspartic-acid-proteases), is one of the initial intracellular biochemical events. The Homogeneous Caspases Assay can be used to detect caspase activity in life-science research applications:
  • for sensitive quantification of activated caspases
  • for screening of caspase inhibitors
  • as an automatable one-step assay for high-throughput screening

Features and Benefits

  • Determine a specific stage of apoptosis
  • Detect caspases of human and animal origin
  • Perform high-throughput screening
  • Minimize hands-on time; use this automatable one-step assay

Packaging

1 kit containing 4 components

Principle

The Homogeneous Caspases Assay is a fluorimetric one-step assay for the quantitative in vitro determination of caspase activity in microplates. Cells are cultured in microplates and apoptosis is induced, causing an activation of caspases. After an appropriate incubation period, caspases substrate (DEVD-R110), diluted in incubation buffer, is added and incubated for 1 - 2 hours. During this incubation period, the cells are lysed by the Incubation buffer and the caspases substrate is cleaved in proportion to the amount of activated caspases. The fluorescent dye Rhodamine 110 is released and fluorometrically determined at λmax = 521nm.
The developed fluorochrome is proportional to the concentration of activated caspases, and can be quantified in a calibration curve.
The total assay time is approximately 1.5 hours (excluding cell culturing).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Substrate (DEVD-R110) Stock Solution 500 μM

  • Positive Control 10x concentrated

  • R 110 Standard

  • Incubation Buffer


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service