Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

11758241001

Roche

β-Gal Reporter Gene Assay, chemiluminescent

sufficient for 500 assays (microplate), sufficient for 250 assays (tubes)

Synonym(s):

galactosidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

usage

sufficient for 250 assays (tubes)
sufficient for 500 assays (microplate)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

The β-Gal Reporter Gene Assay is designed for specifically measuring bacterial β-galactosidase activity. To achieve this, the enzyme reaction is conducted at a pH that is optimized for bacterial β-Gal, but allows no significant endogenous eukaryotic β-galactosidase activity. However, if very high endogenous β-galactosidase activity is found (e.g., with hepatic cells), heat treatment can be performed to ensure the specific determination of the amount of bacterial β-galactosidase that is encoded by the transfected plasmid.

Application

The β-Gal Reporter Gene Assay, chemiluminescent, is used to quantitatively measure β-Gal expression in eukaryotic cells that are transfected with a plasmid bearing the β-Gal-encoding lacZ reporter gene.

Features and Benefits

  • Sensitive: Chemiluminescence technology allows detection of approximately 20 fg β-galactosidase in cell extracts
  • High dynamic measuring range: Linear range over four to five orders of magnitude
  • Constant light emission: The assay produces a long-lasting light emission instead of a short-peak kinetic
  • Safe: No radioactive isotopes are used
  • Fast: Approximately 1.5 to 2.5 hours elapse from start to finish
  • Convenient: Kit contains all reagents needed, including lysis buffer, protease inhibitors, and a positive control

Packaging

1 kit containing 7 components.

Preparation Note

Storage conditions (working solution): Substrate Reagent (solution 1) is stable at least for 24 h at 2 to 8 °C.
Initation Reagent (solution 2) is stable for at least 4 weeks when stored at 2 to 8 °C.
Lysis Reagent, 1x (solution 3) Lysis reagent without protease inhibitors is stable at
-15 to -25 °C or for 3 months at 2 to 8 °C. Lysis reagent containing protease inhibitors is stable for one week when stored frozen at -15 to -25 °C.
Positive Control (solution 4) The reconstituted enzyme solution is stable for 1 week at 2 to 8 °C. When stored frozen in aliquots at -15 to -25 °C the enzyme is stable for 3 months. Avoid multiple freezing and thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • β-Gal Substrate, Galacton Plus 100x concentrated

  • Assay Buffer

  • Enhancer

  • Initiation Solution

  • Protease Inhibitors

  • Lysis Buffer 5x concentrated

  • Positive Control, β-galactosidase (E. coli), lyophilizate

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Analysis of the HOXA2-Dependent Degradation of RCHY1
Bridoux L, et al.
PLoS ONE (2015)
Alexandra Tauzin et al.
Journal of virology, 95(15), e0231120-e0231120 (2021-05-14)
Type I interferons (IFNs) are a family of cytokines that represent a first line of defense against virus infections. The 12 different IFN-α subtypes share a receptor on target cells and trigger similar signaling cascades. Several studies have collectively shown
HOXA2 activity regulation by cytoplasmic relocation, protein stabilization
and post-translational modification
Deneyer, et al.
Biochimica et Biophysica Acta (BBA)-Proteins and Proteomics (2019)
The O-GlcNAc transferase OGT interacts with and post-translationally modifies the transcription factor HOXA1
Draime et al.
Febs Letters (2018)
Arnaud Taminiau et al.
Nucleic acids research, 44(15), 7331-7349 (2016-07-07)
HOX proteins define a family of key transcription factors regulating animal embryogenesis. HOX genes have also been linked to oncogenesis and HOXA1 has been described to be active in several cancers, including breast cancer. Through a proteome-wide interaction screening, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service