Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11585584001

Roche

High Prime DNA Labeling Kit

sufficient for 50 labeling reactions (0.01 to 2 μg DNA per assay)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

usage

sufficient for 50 labeling reactions (0.01 to 2 μg DNA per assay)

manufacturer/tradename

Roche

storage temp.

−20°C

General description

Sample Materials
  • Either linear or supercoiled plasmid DNA, λDNA
  • Shorter fragments of 200 bp
  • DNA fragments in molten agarose
  • <10 ng DNA

Assay Time: 20 minutes for a standard labeling assay

Note: The length of DNA to be labeled does not influence the reaction. Maximal incorporation is achieved after incubation of 30 to 60 minutes.
Convenient kit for the radioactive and nonradioactive labeling of DNA with modified deoxyribonucleoside triphosphate using random oligonucleotides as primers. In this method, the complementary DNA strand of denatured DNA is synthesized by Klenow Polymerase using the 3′-OH termini of the random oligonucleotides as primers.

Specificity

Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0) and/or by heating to 65 °C for 10 minutes.
The standard assay routinely yields a specific activity of 2 x 109 dpm/μg, using different substrate DNAs after 10 minutes of incubation.

Application

Based on the High Prime principle, the High Prime DNA Labeling Kit is used for rapid random-primed DNA labeling. The method enables the labeling of DNAs available only in minute quantities, for example, DNA restriction fragments isolated from gels or in molten agarose. The separately provided nucleotide solutions offer a choice in the selection of modified deoxyribonucleoside triphosphates (e.g., 32P, 35S, 3H, digoxigenin, fluorescein, biotin, or rhodamine). High Prime DNA Labeling Kit has been used for:
  • high prime labeled probes are used in a variety of hybridization techniques: Southern blots
  • northern blots
  • screening of gene libraries
  • in situ hybridizations
  • subtractive hybridization (SSH) assay
The High Prime DNA Labeling Kit offers the same characteristics as High Prime.

Packaging

1 kit containing 6 components.

Principle

The "random primed" DNA labeling method originally developed by Feinberg and Vogelstein is based on the hybridization of oligonucleotides of all possible sequences to the denatured DNA to be labeled.Input DNA serves is the only template for synthesis of labeled DNA, making it possible to label minimal amounts of DNA (10 ng) using this method. Complementary DNA strands are synthesized using Klenow polymerase at the 3′-OH termini of randomized oligonucleotides used as primers. Modified deoxyribonucleoside triphosphates (e.g., labeled with 32P, 35S, 3H, digoxigenin, biotin, fluorescein, or rhodamine) added to the reaction are readily incorporated into newly synthesized DNA strands.

Preparation Note

Working solution: dATP, dGTP, dTTP mixture:
For one labeling reaction pipette:
1 μl dATP, (vial 2)
1 μl dGTP, (vial 4)
1 μl dTTP, (vial 5)
to a reaction vial.

Note: If the same type of labeled deoxyribonucleoside-triphosphate is used repeatedly, we recommend the preparation of a mixture of equal parts of the other three deoxyribonucleoside-triphosphates for convenience.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • High Prime Reaction Mixture (random primer mixture, Klenow polymerase, labeling grade, and 5x stabilized reaction buffer in 50% (v/v) glycerol) 5x concentrated

  • dATP: 2′-Deoxyadenosine-5′-triphosphate in Tris buffer 0.5 mM

  • dCTP: 2′-Deoxycytidine-5′-triphosphate in Tris buffer 0.5 mM

  • dGTP: 2′-Deoxyguanosine-5′-triphosphate in Tris buffer 0.5 mM

  • dTTP: 2′-Deoxy-thymidine-5′-triphosphate in Tris buffer 0.5 mM

  • Control DNA: λDNA 12.5 μg/ml

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Riedl et al.
Journal of immunology (Baltimore, Md. : 1950), 183(1), 370-380 (2009-06-23)
Immunodominance limits the TCR diversity of specific antiviral CD8 T cell responses elicited by vaccination or infection. To prime multispecific T cell responses, we constructed DNA vaccines that coexpress chimeric, multidomain Ags (with CD8 T cell-defined epitopes of the hepatitis
Jia-Cheng Liu et al.
Nucleic acids research, 48(22), 12792-12803 (2020-12-04)
Telomeres at the ends of eukaryotic chromosomes are essential for genome integrality and stability. In order to identify genes that sustain telomere maintenance independently of telomerase recruitment, we have exploited the phenotype of over-long telomeres in the cells that express

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service