Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

10481238001

Roche

pBR322 DNA

solution, pkg of 200 μL (250 μg/ml)

Synonym(s):

dna, pbr 322

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610

form

solution

packaging

pkg of 200 μL (250 μg/ml)

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

storage temp.

−20°C (−15°C to −25°C)

General description

Plasmid for cloning of double-digested restriction fragments.

Application

Select for recombinant DNA. The Pst I and Pvu I cleavage sites in the ampicillin gene, or the Cla I, Hind III, BamH I, and Sal I sites in the tetracycline gene allow the insertion of foreign DNA fragments, and inactivation of one of these genes. pBR322 DNA has been used in to study artificial metallonuclease activity.[1]

Sequence

Chain Length 4,361 bp

Physical form

Solution, 250 μg/ml, in 10 mM Tris-HCl, 1 mM EDTA, pH 8.0, ready-to-use

Storage and Stability

Store at -15 to -25°C until use; once thawed, store at 2-8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andreea Prisecaru et al.
Journal of medicinal chemistry, 56(21), 8599-8615 (2013-10-18)
The synthetic chemical nuclease, [Cu(1,10-phenanthroline)2](2+), has stimulated research within metallonuclease development and in the area of cytotoxic metallodrug design. Our analysis reveals, however, that this agent is "promiscuous" as it binds both dsDNA and protein biomolecules, without specificity, and induces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service