Skip to Content
MilliporeSigma
All Photos(1)

Documents

PRIH3239D258

Princeton BioMolecular Research, Inc.

4-{[(6-oxo-7,8,9,10-tetrahydro-6H-benzo[c]chromen-3-yl)oxy]methyl}benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
350.37
MDL number:

Iupac Name

4-{[(6-oxo-7,8,9,10-tetrahydro-6H-benzo[c]chromen-3-yl)oxy]methyl}benzoic acid

Inchi Code

1S/C21H18O5/c22-20(23)14-7-5-13(6-8-14)12-25-15-9-10-17-16-3-1-2-4-18(16)21(24)26-19(17)11-15/h5-11H,1-4,12H2,(H,22,23)

InChI key

SEQSAFGFIWCZNA-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Product Link

http://store.princetonbio.com/?product_id=PBMR346776

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service