Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ZX0010

Supelco

Zinc 20 Mesh

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Zinc 20 Mesh, Granular Zinc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
EC Index Number:
231-175-3
Assay:
≥99.8% (complexometric)
form:
granular

Agency

meets USP/NF

Assay

≥99.8% (complexometric)

form

granular

color

silver

bp

906 °C/1013 hPa

mp

419 °C

cation traces

Fe: ≤0.01%
Pb: ≤0.01%

shipped in

ambient

storage temp.

room temp

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (complexometric): 99.8% min
Color: Silver metallic
Form: Granular
Iron (Fe): 0.01% max
Lead (Pb): 0.01% max
Mesh size (18-40): 70% min
Suitability for determination of arsenic (As): To pass test (limit 0.1 ppm)

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service