ZBITANK01
Storage Tank
100 L underbench polyethylene storage tank, Protects stored pure (Type 2/3) water
Synonym(s):
100 Liter Polyethylene Underbench Storage Tank
Select a Size
Select a Size
About This Item
Recommended Products
material
polyethylene reservoir
feature
Store and protect pure water:
Underbench integration
packaging
pkg of 1 unit
parameter
98.5 kg operating weight (217.4 lb)
W × D
42 cm (16.6 in.) × 58 cm (22.9 in.)
capacity
100 L
volume
100 L
100000 mL
compatibility
for use with AFS®
for use with Direct-Q® UV
for use with Direct-Q®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® Direct
for use with RiOs™ Essential
for use with RiOs™
Looking for similar products? Visit Product Comparison Guide
General description
The reservoir stores pure water delivered by Elix® Essential/Essential UV/Advantage/Reference, Milli-Q® Direct, Direct-Q®, Direct-Q® UV, RiOs™, RiOs™ Essential, AFS® D, and AFS® Essential water purification systems.
Application
Features and Benefits
- Compact, space-saving design.
- Incorporates the latest technical developments and advanced features for storing water with consistent purity.
- Polyethylene is selected for its minimum release of extractables.
- Opaque walls block sunlight to prevent algae development.
- Smooth inner surface prevents biofilm formation.
- Cylindrical shape minimizes surface area in contact with water.
- Conical bottom allows complete draining for cleaning and rinsing.
- Pure water smoothly fed in at the bottom of the tank prevents the absorption of carbon dioxide.
- Front valve enables manual dispense of pure water.
- Distribution valves permit connection to other laboratory equipment.
- Hermetically sealed lid blocks air from entering the tank.
- Large top opening allows manual cleaning during sanitization procedure.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service