Skip to Content
MilliporeSigma
All Photos(1)

Documents

YY2004040

Millipore

Cartridge Housing Sanitary Clamp 3-tube 2 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161525
eCl@ss:
32031603

diam.

7.5 mm (2 in.)

Quality Level

compatibility

for use with Series 2000 Sanitary Cartridge Housings/3-tube

Related Categories


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service