Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YT3009014

Millipore

Quick-disconnect Male Swagelok

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$79.50

$79.50


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$79.50

About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

$79.50


Please contact Customer Service for Availability

material

316 stainless steel

Quality Level

fitting

1/4 in. inlet/outlet tube

compatibility

for use with ZHE Hazardous Waste Filtration System

shipped in

ambient

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service