Key Documents
XXSPTFF01
Millipore
Pellicon® Single-Pass TFF Mini Kit
Select a Size
XXSPTFF01
$2,553.00
$2,553.00
Please contact Customer Service for Availability
Select a Size
Change View
XXSPTFF01
$2,553.00
About This Item
UNSPSC Code:
23151800
eCl@ss:
32031690
$2,553.00
Please contact Customer Service for Availability
technique(s)
tangential flow filtration (TFF): suitable
Quality Level
General description
The Pellicon® Single-Pass TFF Mini Kit provides 4 diverter plates and 2 silicone gaskets to configure Pellicon® 3 88 cm2, Pellicon® 3 0.11 m² or Pellicon® 2 0.1 m2 cassettes for single-pass TFF operation.
Application
Increase product concentration and recovery. Run intensified, connected or continuous processes.
Legal Information
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany