Skip to Content
MilliporeSigma
All Photos(1)

Documents

XX6310003

Millipore

Small Carrying Bag

Microbiological analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104424
NACRES:
NB.11

Quality Level

input

sample type beer
sample type pharmaceutical(s)
sample type bottled water
sample type cosmetics

application(s)

cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

Incubate total and fecal coliforms, fecal streptococci and other microorganisms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service